siRNA Details
| Gene Symbol: C6orf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014070.2 | NP_054789.2 | 2 | 985 | ||
| GenBank mRNA | AB031481.1 | - | 2 | 985 | ||
| AY358438.1 | - | 2 | 989 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C6orf15| Gene Symbol: C6orf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014070.2 | NP_054789.2 | 2 | 985 | ||
| GenBank mRNA | AB031481.1 | - | 2 | 985 | ||
| AY358438.1 | - | 2 | 989 | |||