siRNA Details
| Gene Symbol: CCDC106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NM_013301.2 | NP_037433.2 | 3 | 828 | ||
| XM_005258827.2 | XP_005258884.1 | 3 | 426 | |||
| XM_005258828.2 | XP_005258885.1 | 3 | 447 | |||
| GenBank mRNA | AF054984.1 | - | 3 | 381 | ||
| AK097095.1 | - | 2 | 998 | |||
| AY927563.1 | - | 1 | 363 | |||
| AY927628.1 | - | LIT3614 AY927628 |
2 | 236 | ||
| BC000412.2 | - | 2 | 406 | 2961290 | ||
| BC008956.2 | - | 2 | 406 | 2961290 | ||
| CR450308.1 | - | 2 | 93 | |||
| GU354321.1 | - | 3 | 183 | |||
| GU354322.1 | - | 3 | 239 | |||
| GU354323.1 | - | 3 | 427 | |||
| U79303.1 | - | 3 | 381 | |||

CCDC106
Genomic Map