siRNA Details
| Gene Symbol: PRLH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015893.1 | NP_056977.1 | 2 | 120 | ||
| GenBank mRNA | AB015419.1 | - | 2 | 120 | ||
| BC069081.1 | - | 2 | 120 | 7216895 | ||
| BC069284.1 | - | 2 | 120 | 7262679 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRLH| Gene Symbol: PRLH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015893.1 | NP_056977.1 | 2 | 120 | ||
| GenBank mRNA | AB015419.1 | - | 2 | 120 | ||
| BC069081.1 | - | 2 | 120 | 7216895 | ||
| BC069284.1 | - | 2 | 120 | 7262679 | ||