siRNA Details
| Gene Symbol: ZNF580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163423.1 | NP_001156895.1 | 2 | 769 | ||
| NM_016202.2 | NP_057286.1 | 1 | 1162 | |||
| NM_207115.1 | NP_996998.1 | 2 | 875 | |||
| GenBank mRNA | AF184939.1 | - | 1 | 1449 | ||
| AL359054.1 | - | 2 | 965 | |||
| BC017698.1 | - | 1 | 959 | 4431902 | ||

ZNF580
Genomic Map