siRNA Details
Gene Symbol: MUC3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005960.1 | NP_005951.1 | 11, 12 | 10130 | ||
XM_011516246.2 | XP_011514548.1 | 12, 13 | 10459 | |||
XM_011516247.2 | XP_011514549.1 | 12, 13 | 10409 | |||
XM_011516249.2 | XP_011514551.1 | 12, 13 | 10260 | |||
XM_011516250.2 | XP_011514552.1 | 12, 13 | 9748 | |||
XM_017012231.1 | XP_016867720.1 | 12, 13 | 9956 | |||
GenBank mRNA | AB038784.1 | - | 10, 11 | 2587 | ||
AF143371.1 | - | 9, 10 | 1054 | |||
AF143372.1 | - | 8, 9 | 880 | |||
AF143373.1 | - | 8, 9 | 938 |
Gene Symbol: MUC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB038783.1 | - | 10, 11 | 2590 |
