siRNA Details
| Gene Symbol: C19orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317801.1 | NP_001304730.1 | 3, 4 | 361 | ||
| NM_033520.2 | NP_277055.1 | 3, 4 | 302 | |||
| GenBank mRNA | AB038317.1 | - | 3, 4 | 299 | ||
| AB038318.1 | - | 3, 4 | 358 | |||
| AF213678.1 | - | 3, 4 | 302 | |||
| BC008888.2 | - | 3, 4 | 259 | |||
| BC014264.2 | - | 3, 4 | 259 | 4550805 | ||
| BC060319.1 | - | 3, 4 | 313 | 3901226 | ||
| BX648823.1 | - | 3, 4 | 263 | |||
| CB109881.1 | - | 3, 4 | 207 | |||

C19orf33
Genomic Map