siRNA Details
| Gene Symbol: RABEP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024816.2 | NP_079092.2 | 4 | 574 | ||
| GenBank mRNA | AK026935.1 | - | 4 | 574 | ||
| AK092299.1 | - | 4 | 1115 | |||
| AK295229.1 | - | 3 | 346 | |||
| BC027584.1 | - | 4 | 1052 | 4827815 | ||
| BC046118.1 | - | 4 | 568 | |||
| BC058900.1 | - | 4 | 577 | 5415624 | ||

RABEP2
Genomic Map