siRNA Details
| Gene Symbol: BAIAP2L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025045.5 | NP_079321.3 | 6 | 582 | ||
| XM_005261751.4 | XP_005261808.1 | 7 | 583 | |||
| XM_011530379.2 | XP_011528681.1 | 6 | 699 | |||
| XM_011530380.2 | XP_011528682.1 | 5 | 485 | |||
| XM_011530381.2 | XP_011528683.1 | 6 | 476 | |||
| XM_011530382.2 | XP_011528684.1 | 6 | 864 | |||
| XM_011530383.2 | XP_011528685.1 | 5 | 802 | |||
| XM_011530384.2 | XP_011528686.1 | 6 | 740 | |||
| GenBank mRNA | AK026235.1 | - | 6 | 540 | ||
| AY358164.1 | - | 6 | 582 | |||

BAIAP2L2
Genomic Map