siRNA Details
| Gene Symbol: UPK3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030570.3 | NP_085047.1 | 1 | 503 | ||
| NM_182683.2 | NP_872624.1 | 2 | 338 | |||
| NM_182684.2 | NP_872625.1 | 2 | 338 | |||
| GenBank mRNA | AK056760.1 | - | 2 | 146 | ||
| AK290096.1 | - | 1 | 406 | |||
| AY233462.1 | - | 2 | 171 | |||
| BC004304.1 | - | 1 | 327 | 3625247 | ||
| BE615406.1 | - | 2 | 337 | |||
| BM921749.1 | - | 362 | ||||
| BP352210.1 | - | 1 | 565 | |||
| BQ689127.1 | - | 333 | ||||
| BU150542.1 | - | 333 | ||||
| Gene Symbol: LOC105375355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005250612.3 | XP_005250669.1 | 1 | 824 | ||
| XM_005250613.4 | XP_005250670.2 | 2 | 416 | |||
| XM_005250614.2 | XP_005250671.1 | 2 | 416 | |||
| XM_006716140.3 | XP_006716203.1 | 2 | 416 | |||
| XM_006716141.3 | XP_006716204.1 | 2 | 416 | |||
| XM_017012885.1 | XP_016868374.1 | 2 | 416 | |||
| XM_017012886.1 | XP_016868375.1 | 2 | 416 | |||

UPK3B
Genomic Map