siRNA Details
| Gene Symbol: MIXL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282402.1 | NP_001269331.1 | 2 | 562 | ||
| NM_031944.2 | NP_114150.1 | 2 | 538 | |||
| GenBank mRNA | AF211891.1 | - | 2 | 474 | ||
| BC111974.1 | - | 2 | 538 | 8327442 | ||
| BC113441.1 | - | 2 | 538 | 8322493 | ||
| BC143784.1 | - | 2 | 562 | 9052298 | ||

MIXL1
Genomic Map