siRNA Details
| Gene Symbol: RNASEH2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032193.3 | NP_115569.2 | 4 | 842 | ||
| GenBank mRNA | AK225131.1 | - | 4 | 699 | ||
| AK297660.1 | - | 4 | 612 | |||
| BM921525.1 | - | 4 | 711 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNASEH2C| Gene Symbol: RNASEH2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032193.3 | NP_115569.2 | 4 | 842 | ||
| GenBank mRNA | AK225131.1 | - | 4 | 699 | ||
| AK297660.1 | - | 4 | 612 | |||
| BM921525.1 | - | 4 | 711 | |||