siRNA Details
| Gene Symbol: ZNF414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032370.2 | NP_115746.2 | 6 | 1161 | ||
| GenBank mRNA | BC007310.1 | - | 6 | 1145 | 3353224 | |
| BC071937.1 | - | 6 | 1140 | 5089259 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF414| Gene Symbol: ZNF414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032370.2 | NP_115746.2 | 6 | 1161 | ||
| GenBank mRNA | BC007310.1 | - | 6 | 1145 | 3353224 | |
| BC071937.1 | - | 6 | 1140 | 5089259 | ||