siRNA Details
| Gene Symbol: PARP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317895.1 | NP_001304824.1 | 5 | 1733 | ||
| NM_032789.4 | NP_116178.2 | 6 | 1609 | |||
| NR_134234.1 | 6 | 1609 | ||||
| XM_011517336.2 | XP_011515638.1 | 6 | 1576 | |||
| GenBank mRNA | AK027370.1 | - | 6 | 1566 | ||
| AK075250.1 | - | 2 | 296 | |||
| AK128537.1 | - | 7 | 1424 | |||
| AK129821.1 | - | 2 | 114 | |||
| AK222914.1 | - | 6 | 1592 | |||
| AK303317.1 | - | 5 | 1658 | |||
| BC019030.2 | - | 2 | 429 | |||
| BC020378.1 | - | 6 | 1584 | |||
| BC030549.1 | - | 6 | 1583 | |||
| BC132961.1 | - | 6 | 1560 | 40146983 | ||
| BC136542.1 | - | 6 | 1560 | 9020532 | ||
| BC144235.1 | - | 6 | 1560 | |||

PARP10
Genomic Map