siRNA Details
| Gene Symbol: ZDHHC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318015.1 | NP_001304944.1 | 2 | 391 | ||
| NM_001318016.1 | NP_001304945.1 | 2 | 226 | |||
| NM_001318020.1 | NP_001304949.1 | 2 | 226 | |||
| NM_001318023.1 | NP_001304952.1 | 2 | 226 | |||
| NM_032799.4 | NP_116188.2 | 2 | 226 | |||
| XM_011519116.2 | XP_011517418.1 | 2 | 400 | |||
| GenBank mRNA | AK027430.1 | - | 2 | 219 | ||
| AK075332.1 | - | 2 | 219 | |||
| AK092843.1 | - | 1 | 1643 | |||
| AK315746.1 | - | 2 | 222 | |||
| BC009280.2 | - | 2 | 203 | 4302257 | ||
| BC048251.1 | - | 2 | 372 | 5418875 | ||
| BG767826.1 | - | 2 | 200 | |||
| BM043458.1 | - | 2 | 194 | |||
| BM745076.1 | - | 2 | 218 | |||

ZDHHC12
Genomic Map