siRNA Details
| Gene Symbol: BAGE4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181704.1 | NP_859055.1 | 372 | |||
| GenBank mRNA | AF339515.1 | - | 372 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BAGE4| Gene Symbol: BAGE4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181704.1 | NP_859055.1 | 372 | |||
| GenBank mRNA | AF339515.1 | - | 372 | |||