siRNA Details
Gene Symbol: DGCR6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033257.3 | NP_150282.2 | 5 | 798 | ||
GenBank mRNA | AF228708.1 | - | 5 | 778 | ||
AK289952.1 | - | 5 | 716 | |||
AK307210.1 | - | 4 | 633 | |||
AK310899.1 | - | 4 | 1284 | |||
BC000682.2 | - | 5 | 734 | 3349309 | ||
CR456361.1 | - | 5 | 703 |
Gene Symbol: DGCR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005675.4 | NP_005666.2 | 5 | 833 | ||
XM_011530394.1 | XP_011528696.1 | 6 | 820 | |||
GenBank mRNA | AF228707.1 | - | 5 | 777 | ||
AK098780.1 | - | 5 | 1055 | |||
BC047039.1 | - | 5 | 726 | 5229172 | ||
CR456434.1 | - | 5 | 703 | |||
X96484.1 | - | 5 | 695 |
Gene Symbol: LOC102724770 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724934.1 | XP_006724997.1 | 5 | 775 |
