siRNA Details
| Gene Symbol: MIDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177401.4 | NP_796375.3 | 6, 7 | 1369 | ||
| XM_005259671.3 | XP_005259728.1 | 7, 8 | 1498 | |||
| XM_005259672.3 | XP_005259729.1 | 7, 8 | 1496 | |||
| GenBank mRNA | BC094778.1 | - | 5, 6 | 1263 | 30342357 | |

MIDN
Genomic Map