siRNA Details
| Gene Symbol: FAM181A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001207071.1 | NP_001194000.1 | 2 | 1431 | ||
| NM_001207072.1 | NP_001194001.1 | 2 | 1572 | |||
| NM_001207073.1 | NP_001194002.1 | 2 | 1442 | |||
| NM_001207074.1 | NP_001194003.1 | 2 | 1388 | |||
| NM_138344.4 | NP_612353.3 | 3 | 1754 | |||
| XM_011537297.1 | XP_011535599.1 | 2 | 1405 | |||
| GenBank mRNA | AK093387.1 | - | 3 | 1739 | ||
| BC009073.1 | - | 2 | 1572 | 3856004 | ||

FAM181A
Genomic Map