siRNA Details
| Gene Symbol: ZNF251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138367.1 | NP_612376.1 | 4 | 510 | ||
| XM_005272356.4 | XP_005272413.1 | 4 | 528 | |||
| XM_006716683.2 | XP_006716746.1 | 3 | 329 | |||
| XM_011517361.1 | XP_011515663.1 | 3 | 329 | |||
| GenBank mRNA | AK000435.1 | - | 4 | 394 | ||
| AK091638.1 | - | 2 | 2317 | |||
| DA811573.1 | - | 4 | 510 | |||

ZNF251
Genomic Map