siRNA Details
| Gene Symbol: HIST3H2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033445.2 | NP_254280.1 | 1 | 158 | ||
| GenBank mRNA | AK311930.1 | - | 1 | 170 | ||
| BC001193.1 | - | 1 | 158 | |||
| BC082269.1 | - | 1 | 148 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIST3H2A| Gene Symbol: HIST3H2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033445.2 | NP_254280.1 | 1 | 158 | ||
| GenBank mRNA | AK311930.1 | - | 1 | 170 | ||
| BC001193.1 | - | 1 | 158 | |||
| BC082269.1 | - | 1 | 148 | |||