siRNA Details
| Gene Symbol: KCNG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172347.2 | NP_758857.1 | 2 | 171 | ||
| GenBank mRNA | AF348984.1 | - | 1 | 50 | ||
| AF450108.1 | - | 1 | 50 | |||
| BC008969.2 | - | 2 | 171 | |||
| BC109203.2 | - | 2 | 119 | 40005993 | ||

KCNG4
Genomic Map