siRNA Details
| Gene Symbol: TUBA3D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080386.3 | NP_525125.2 | 3 | 537 | ||
| GenBank mRNA | AK093116.1 | - | 3 | 451 | ||
| AK292253.1 | - | 3 | 424 | |||
| BC057810.1 | - | 3 | 423 | 5268074 | ||
| DB051220.1 | - | 3 | 540 | |||
| Gene Symbol: TUBA3E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207312.2 | NP_997195.1 | 3 | 445 | ||
| GenBank mRNA | AK131043.1 | - | 3 | 423 | ||
| BC057811.1 | - | 3 | 445 | 5298532 | ||

TUBA3D
Genomic Map