siRNA Details
| Gene Symbol: MFSD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138431.2 | NP_612440.1 | 1, 2 | 977 | ||
| XM_017013005.1 | XP_016868494.1 | 1, 2 | 1932 | |||
| GenBank mRNA | BC011982.1 | - | 1, 2 | 844 | 3860434 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

MFSD3| Gene Symbol: MFSD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138431.2 | NP_612440.1 | 1, 2 | 977 | ||
| XM_017013005.1 | XP_016868494.1 | 1, 2 | 1932 | |||
| GenBank mRNA | BC011982.1 | - | 1, 2 | 844 | 3860434 | |