siRNA Details
| Gene Symbol: LRRC56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198075.3 | NP_932341.1 | 14 | 2711 | ||
| XM_005252775.2 | XP_005252832.1 | 14 | 3535 | |||
| XM_005252776.2 | XP_005252833.1 | 13 | 3358 | |||
| XM_006718132.2 | XP_006718195.1 | 14 | 3610 | |||
| XM_006718133.2 | XP_006718196.1 | 13 | 2418 | |||
| XM_011519875.2 | XP_011518177.1 | 15 | 2950 | |||
| XM_011519876.1 | XP_011518178.1 | 15 | 3606 | |||
| XM_011519877.2 | XP_011518179.1 | 14 | 2687 | |||
| XM_017017167.1 | XP_016872656.1 | 15 | 3025 | |||
| XM_017017168.1 | XP_016872657.1 | 15 | 3264 | |||
| GenBank mRNA | AK024495.1 | - | 11 | 4601 | ||
| AL831961.1 | - | 3 | 1077 | |||
| BC016439.1 | - | 12 | 2245 | 3447051 | ||
| BC035937.1 | - | 14 | 2697 | 4816105 | ||

LRRC56
Genomic Map