siRNA Details
| Gene Symbol: WFIKKN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053284.2 | NP_444514.1 | 2 | 663 | ||
| GenBank mRNA | AF422194.1 | - | 2 | 421 | ||
| AK075356.1 | - | 2 | 743 | |||
| AK128777.1 | - | 1 | 3591 | |||
| AK314757.1 | - | 2 | 725 | |||
| BC101602.1 | - | 2 | 504 | 8069108 | ||
| BC101606.1 | - | 2 | 504 | 8069112 | ||

WFIKKN1
Genomic Map