siRNA Details
Gene Symbol: TMEM219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083613.1 | NP_001077082.1 | 4 | 512 | ||
NM_194280.3 | NP_919256.1 | 4 | 509 | |||
XM_005255123.1 | XP_005255180.1 | 4 | 511 | |||
GenBank mRNA | BC017488.1 | - | 3 | 277 | ||
BC031217.2 | - | 3 | 406 | |||
BC050051.1 | - | 4 | 539 | 5278133 | ||
BF724791.1 | - | 4 | 469 | |||
CF453990.1 | - | 456 | ||||
FJ748884.1 | - | 3 | 372 |
