siRNA Details
| Gene Symbol: C19orf70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308240.1 | NP_001295169.1 | 4, 5 | 870 | ||
| NM_205767.2 | NP_991330.1 | 3, 4 | 674 | |||
| XM_017026248.1 | XP_016881737.1 | 3, 4 | 615 | |||
| GenBank mRNA | AA292888.1 | - | 3, 4 | 441 | ||
| AJ707830.1 | - | 3, 4 | 420 | |||
| BC009557.1 | - | 3, 4 | 308 | 3899541 | ||
| BC042386.1 | - | 3, 4 | 283 | 5288954 | ||
| BC052640.1 | - | 3, 4 | 285 | |||
| BI827162.1 | - | 3, 4 | 673 | |||

C19orf70
Genomic Map