siRNA Details
| Gene Symbol: SPATA25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080608.3 | NP_542175.1 | 2 | 698 | ||
| GenBank mRNA | BC039607.1 | - | 2 | 698 | 5167905 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPATA25| Gene Symbol: SPATA25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080608.3 | NP_542175.1 | 2 | 698 | ||
| GenBank mRNA | BC039607.1 | - | 2 | 698 | 5167905 | |