siRNA Details
| Gene Symbol: BANF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014977.3 | NP_001014977.2 | 3 | 386 | ||
| NM_001159495.1 | NP_001152967.1 | 3 | 310 | |||
| NM_178477.4 | NP_848572.3 | 4 | 549 | |||
| XM_005260668.3 | XP_005260725.1 | 5 | 922 | |||
| XM_011529170.1 | XP_011527472.1 | 4 | 759 | |||
| XM_011529171.2 | XP_011527473.1 | 4 | 561 | |||
| GenBank mRNA | AA394107.1 | - | 3 | 349 | ||
| AW022487.1 | - | 3 | 318 | |||
| BC054871.1 | - | 3 | 390 | 6592641 | ||
| BC062735.1 | - | 4 | 540 | 6591888 | ||

BANF2
Genomic Map