siRNA Details
| Gene Symbol: SLC22A31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242757.1 | NP_001229686.1 | 4 | 792 | ||
| XM_011522897.2 | XP_011521199.1 | 5 | 840 | |||
| XM_011522899.2 | XP_011521201.1 | 5 | 777 | |||
| XM_017022962.1 | XP_016878451.1 | 2 | 442 | |||
| XM_017022963.1 | XP_016878452.1 | 2 | 2190 | |||
| XM_017022964.1 | XP_016878453.1 | 3 | 1346 | |||
| XM_017022965.1 | XP_016878454.1 | 2 | 2131 | |||
| XM_017022966.1 | XP_016878455.1 | 3 | 1969 | |||

SLC22A31
Genomic Map