siRNA Details
| Gene Symbol: XAGE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130776.1 | NP_570132.1 | 4, 5 | 379 | ||
| NM_133179.2 | NP_573440.1 | 4, 5 | 410 | |||
| XM_011530768.2 | XP_011529070.1 | 4, 5 | 476 | |||
| XM_011530769.2 | XP_011529071.1 | 4, 5 | 439 | |||
| XM_011530770.2 | XP_011529072.1 | 4, 5 | 403 | |||
| GenBank mRNA | AF318372.1 | - | 4, 5 | 391 | ||
| AJ318881.1 | - | 4, 5 | 379 | |||
| BC062680.1 | - | 4, 5 | 414 | 4619720 | ||

XAGE3
Genomic Map