siRNA Details
| Gene Symbol: XAGE5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130775.2 | NP_570131.1 | 5 | 428 | ||
| XM_017029320.1 | XP_016884809.1 | 6 | 661 | |||
| GenBank mRNA | BC069129.1 | - | 5 | 429 | 7216985 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

XAGE5| Gene Symbol: XAGE5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130775.2 | NP_570131.1 | 5 | 428 | ||
| XM_017029320.1 | XP_016884809.1 | 6 | 661 | |||
| GenBank mRNA | BC069129.1 | - | 5 | 429 | 7216985 | |