siRNA Details
| Gene Symbol: EME2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257370.1 | NP_001244299.1 | 7 | 895 | ||
| XM_017023025.1 | XP_016878514.1 | 7 | 817 | |||
| XM_017023026.1 | XP_016878515.1 | 6 | 808 | |||
| GenBank mRNA | AK074080.2 | - | 6 | 979 | ||
| EF452422.1 | - | 7 | 895 | |||

EME2
Genomic Map