siRNA Details
| Gene Symbol: GGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152657.3 | NP_689870.3 | 3 | 1320 | ||
| XM_005258619.4 | XP_005258676.1 | 3 | 1682 | |||
| XM_011526603.2 | XP_011524905.1 | 3 | 1432 | |||
| XM_017026451.1 | XP_016881940.1 | 2 | 1526 | |||
| GenBank mRNA | AF538035.1 | - | 3 | 1325 | ||
| AK057356.2 | - | 3 | 1060 | |||
| AK093032.1 | - | 2 | 1268 | |||
| BC035008.2 | - | 3 | 1320 | 5269088 | ||

GGN
Genomic Map