siRNA Details
| Gene Symbol: RWDD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320724.1 | NP_001307653.1 | 6 | 1089 | ||
| NM_016940.2 | NP_058636.1 | 5 | 984 | |||
| GenBank mRNA | AF212232.1 | - | 5 | 945 | ||
| AK000056.1 | - | 5 | 988 | |||
| AK223357.1 | - | 5 | 995 | |||
| AL355726.1 | - | 1 | 138 | |||
| BC012546.1 | - | 5 | 952 | 4291536 | ||
| BC017912.1 | - | 5 | 970 | 4291966 | ||
| Y19009.1 | - | 5 | 963 | |||

RWDD2B
Genomic Map