siRNA Details
| Gene Symbol: TMEM211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001663.1 | NP_001001663.1 | 3 | 281 | ||
| XM_011530094.2 | XP_011528396.1 | 2 | 1158 | |||
| GenBank mRNA | BC130647.1 | - | 3 | 280 | 40146678 | |
| BC130649.1 | - | 3 | 280 | 40146680 | ||
| CR456388.1 | - | 3 | 281 | |||

TMEM211
Genomic Map