siRNA Details
| Gene Symbol: TINCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027064.2 | 3 | 1203 | |||
| GenBank mRNA | BC036545.1 | - | 3 | 1170 | ||
| BC120980.1 | - | 2 | 816 | |||
| BC120981.1 | - | 2 | 816 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TINCR| Gene Symbol: TINCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027064.2 | 3 | 1203 | |||
| GenBank mRNA | BC036545.1 | - | 3 | 1170 | ||
| BC120980.1 | - | 2 | 816 | |||
| BC120981.1 | - | 2 | 816 | |||