siRNA Details
Gene Symbol: NPW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099456.2 | NP_001092926.2 | 2 | 974 | ||
GenBank mRNA | BC171886.1 | - | 1 | 1411 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NPW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099456.2 | NP_001092926.2 | 2 | 974 | ||
GenBank mRNA | BC171886.1 | - | 1 | 1411 |