siRNA Details
| Gene Symbol: ODF3L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182577.2 | NP_872383.1 | 4 | 1235 | ||
| XM_005259545.4 | XP_005259602.1 | 4 | 1408 | |||
| XM_017026671.1 | XP_016882160.1 | 3 | 805 | |||
| GenBank mRNA | AK097378.1 | - | 4 | 1234 | ||
| BC104468.1 | - | 3 | 1023 | 40032428 | ||
| BC104469.1 | - | 4 | 1127 | 40032432 | ||

ODF3L2
Genomic Map