siRNA Details
| Gene Symbol: NBPF15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170755.2 | NP_001164226.1 | 12 | 1830 | ||
| NM_173638.4 | NP_775909.2 | 9 | 1300 | |||
| XM_017001077.1 | XP_016856566.1 | 7 | 948 | |||
| GenBank mRNA | AF379630.1 | - | 4 | 478 | ||
| AK097180.1 | - | 12 | 1749 | |||
| AK290302.1 | - | 6 | 759 | |||
| AK299360.1 | - | 3 | 340 | |||
| AY894566.1 | - | 6 | 740 | |||
| BC023087.1 | - | 9 | 1157 | 3918932 | ||
| DC372755.1 | - | 3 | 340 | |||

NBPF15
Genomic Map