siRNA Details
| Gene Symbol: GPR150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199243.1 | NP_954713.1 | 1 | 1039 | ||
| GenBank mRNA | JF810884.1 | - | 1 | 1039 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR150| Gene Symbol: GPR150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199243.1 | NP_954713.1 | 1 | 1039 | ||
| GenBank mRNA | JF810884.1 | - | 1 | 1039 | ||
Set Membership: |
|