siRNA Details
| Gene Symbol: NBPF22P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003719.2 | 3 | 576 | |||
| GenBank mRNA | AK308274.1 | - | 3 | 560 | ||
| BC050328.1 | - | 3 | 576 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NBPF22P| Gene Symbol: NBPF22P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003719.2 | 3 | 576 | |||
| GenBank mRNA | AK308274.1 | - | 3 | 560 | ||
| BC050328.1 | - | 3 | 576 | |||