siRNA Details
| Gene Symbol: HIST2H2AB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175065.2 | NP_778235.1 | 1 | 364 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIST2H2AB| Gene Symbol: HIST2H2AB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175065.2 | NP_778235.1 | 1 | 364 | ||