siRNA Details
| Gene Symbol: KRTAP6-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181604.1 | NP_853635.1 | 1 | 119 | ||
| GenBank mRNA | AB096952.1 | - | 1 | 145 | ||
| BC093915.1 | - | 1 | 147 | |||
| BC093917.1 | - | 1 | 147 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP6-2| Gene Symbol: KRTAP6-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181604.1 | NP_853635.1 | 1 | 119 | ||
| GenBank mRNA | AB096952.1 | - | 1 | 145 | ||
| BC093915.1 | - | 1 | 147 | |||
| BC093917.1 | - | 1 | 147 | |||