siRNA Details
| Gene Symbol: LINC00936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028138.1 | 1 | 568 | |||
| GenBank mRNA | BC040326.1 | - | 1 | 564 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00936| Gene Symbol: LINC00936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028138.1 | 1 | 568 | |||
| GenBank mRNA | BC040326.1 | - | 1 | 564 | ||