siRNA Details
| Gene Symbol: TMEM52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178545.3 | NP_848640.1 | 4 | 359 | ||
| GenBank mRNA | AJ278736.1 | - | 4 | 373 | ||
| AY358544.1 | - | 1 | 519 | |||
| CN263898.1 | - | 4 | 371 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM52| Gene Symbol: TMEM52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178545.3 | NP_848640.1 | 4 | 359 | ||
| GenBank mRNA | AJ278736.1 | - | 4 | 373 | ||
| AY358544.1 | - | 1 | 519 | |||
| CN263898.1 | - | 4 | 371 | |||