siRNA Details
| Gene Symbol: TSSC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297658.1 | NP_001284587.1 | 4 | 931 | ||
| NM_001297659.1 | NP_001284588.1 | 3 | 779 | |||
| NM_001297660.1 | NP_001284589.1 | 4 | 763 | |||
| NM_005706.3 | NP_005697.2 | 3 | 622 | |||
| XM_006718118.2 | XP_006718181.1 | 3 | 1661 | |||
| XM_011519830.2 | XP_011518132.1 | 4 | 873 | |||
| GenBank mRNA | AF125568.1 | - | 2 | 312 | ||
| BC006091.1 | - | 3 | 335 | 3677524 | ||
| BI768923.1 | - | 4 | 656 | |||
| BX384913.2 | - | 4 | 492 | |||

TSSC4
Genomic Map