siRNA Details
| Gene Symbol: SYCN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080468.2 | NP_001073937.1 | 1 | 378 | ||
| GenBank mRNA | AK130951.1 | - | 1 | 389 | ||
| BC121075.2 | - | 1 | 382 | 40119665 | ||
| BC121076.2 | - | 1 | 382 | 40119666 | ||

SYCN
Genomic Map