siRNA Details
| Gene Symbol: ATP9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006045.2 | NP_006036.1 | 24, 25 | 2671 | ||
| XM_011528481.2 | XP_011526783.1 | 24, 25 | 2699 | |||
| GenBank mRNA | AB014511.1 | - | 19, 20 | 2225 | ||
| AK172802.1 | - | 22, 23 | 2360 | |||
| AK299064.1 | - | 24, 25 | 2600 | |||
| BC110591.1 | - | 20, 21 | 2163 | |||
| BC110592.1 | - | 20, 21 | 2163 | |||

ATP9A
Genomic Map