siRNA Details
| Gene Symbol: ATP9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006045.2 | NP_006036.1 | 15 | 1563 | ||
| XM_011528481.2 | XP_011526783.1 | 15 | 1591 | |||
| GenBank mRNA | AB014511.1 | - | 10 | 1117 | ||
| AK172802.1 | - | 13 | 1252 | |||
| AK299064.1 | - | 15 | 1492 | |||
| BC110591.1 | - | 11 | 1055 | |||
| BC110592.1 | - | 11 | 1055 | |||

ATP9A
Genomic Map